Dibutyl vinylboronate ≥94.0% (by GC) stabilised
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Stabilisatie:
Stabilised with phenothiazine
D3596-1G
D3596-5G
TCIAD3596-1GEA
60.5
EUR
TCIAD3596-1G
TCIAD3596-5G
Dibutyl vinylboronate ≥94.0% (by GC) stabilised
Dibutyl vinylboronate
Formule:
C₁₀H₂₁BO₂ Kookpunt: 89°C Vlampunt: 67°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00050049 CAS nummer: 6336-45-4 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...