Diisopropyl(bromomethyl)boronate ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: (Bromomethyl)boronic Acid Diisopropyl Ester

D3649-1G D3649-5G
TCIAD3649-1GEA 114 EUR
TCIAD3649-1G TCIAD3649-5G
Diisopropyl(bromomethyl)boronate ≥95.0% (by GC)
Diisopropyl(bromomethyl)boronate
Formule: C₇H₁₆BBrO₂
MW: 222.92 g/mol
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD01631222
CAS nummer: 137297-49-5
UN: 3272
ADR: 3 III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR