(2S,3S)-(+)-2,3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one ≥96.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D3661-25G D3661-5G
TCIAD3661-25GEA 95.7 EUR
TCIAD3661-25G TCIAD3661-5G
(2S,3S)-(+)-2,3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one ≥96.0% (by HPLC, total nitrogen)
(2S,3S)-(+)-2,3-Dihydro-3-hydroxy-2-(4-methoxyphenyl)-1,5-benzothiazepin-4(5H)-one
Formule: C₁₆H₁₅NO₃S
MDL Nummer: MFCD01321316
CAS nummer: 42399-49-5
EINECS: 255-799-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (with Total Nitrogen) min. 96.0 %
Melting point 201.0 to 207.0 deg-C
Specific rotation [a]20/D 110 to 118 deg (c=0.5, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR