Dimethyl3,4-dihydroxy-2,5-thiophenedicarboxylate ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D3682-1G
D3682-5G
TCIAD3682-1GEA
88
EUR
TCIAD3682-1G
TCIAD3682-5G
Dimethyl3,4-dihydroxy-2,5-thiophenedicarboxylate ≥98.0% (by GC)
Dimethyl3,4-dihydroxy-2,5-thiophenedicarboxylate
Formule:
C₈H₈O₆S |
MDL Nummer:
MFCD01860087 CAS nummer: 58416-04-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 180.0 to 184.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...