1,3-Dioxan-2-one ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Trimethylene Carbonate
D3821-5G
D3821-25G
TCIAD3821-5GEA
193
EUR
TCIAD3821-5G
TCIAD3821-25G
1,3-Dioxan-2-one ≥98.0% (by GC)
1,3-Dioxan-2-one
Formule:
C₄H₆O₃ Kookpunt: 135°C |
MDL Nummer:
MFCD00010440 CAS nummer: 2453-03-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 45.0 to 49.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...