Diisopropyl fumarate ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D3916-25G D3916-500G
TCIAD3916-25GEA 17.6 EUR
TCIAD3916-25G TCIAD3916-500G
Diisopropyl fumarate ≥98.0% (by GC)
Diisopropyl fumarate
Formule: C₁₀H₁₆O₄
Kookpunt: 110°C
Smeltpunt: 2°C
MDL Nummer: MFCD00545172
CAS nummer: 7283-70-7
EINECS: 230-707-1
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %