Disodium pamidronate, hydrat ≥97.0% (by titrimetric analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D3921-1G D3921-100MG
TCIAD3921-1GEA 422 EUR
TCIAD3921-1G TCIAD3921-100MG
Disodium pamidronate, hydrat ≥97.0% (by titrimetric analysis)
Disodium pamidronate
Formule: C₃H₉NNa₂O₇P₂·xH₂O
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD11045314
CAS nummer: 57248-88-1
EINECS: 260-647-1
Merck Index: 14,07003
Order Now
Specification Test Results
Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 % (calcd.on anh.substance)
Water max. 25.0 %