1,4-Dimethyl-1,2,4-triazolium iodide ≥98.0% (by HPLC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
1,4-Dimethyl-1,2,4-triazolium Iodide
D3962-5G
D3962-1G
TCIAD3962-5GEA
198
EUR
TCIAD3962-5G
TCIAD3962-1G
1,4-Dimethyl-1,2,4-triazolium iodide ≥98.0% (by HPLC, titration analysis)
1,4-Dimethyl-1,2,4-triazolium iodide
Formule:
C₄H₈IN₃ |
MDL Nummer:
MFCD00666522 CAS nummer: 120317-69-3 |
|
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) (CAD) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 118.0 to 124.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...