2,2-Dimethylbutyryl chloride ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
D3987-25G D3987-100G
TCIAD3987-25GEA 41.8 EUR
TCIAD3987-25G TCIAD3987-100G
2,2-Dimethylbutyryl chloride ≥98.0% (by GC, titration analysis)
2,2-Dimethylbutyryl chloride
Formule: C₆H₁₁ClO
Kookpunt: 132°C
Vlampunt: 29°C
MDL Nummer: MFCD03093071
CAS nummer: 5856-77-9
EINECS: 227-478-5
UN: 2920
ADR: 8,II
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %