Diethyl-2,5-dibromoterephthalate ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2,5-Dibromoterephthalic Acid Diethyl Ester, Diethyl 2,5-dibromoterephthalate

D3995-25G D3995-5G
TCIAD3995-25GEA 328 EUR
TCIAD3995-25G TCIAD3995-5G
Diethyl-2,5-dibromoterephthalate ≥98.0% (by GC)
Diethyl-2,5-dibromoterephthalate
Formule: C₁₂H₁₂Br₂O₄
MW: 380.03 g/mol
Kookpunt: 335°C
MDL Nummer: MFCD12407163
CAS nummer: 18013-97-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 127.0 to 131.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR