Diacerein ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 4,5-Diacetoxy-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid, 4,5-Diacetoxyanthraquinone-2-carboxylic Acid

D4061-1G D4061-5G
TCIAD4061-1GEA 67.1 EUR
TCIAD4061-1G TCIAD4061-5G
Diacerein ≥98.0% (by HPLC)
Diacerein
Formule: C₁₉H₁₂O₈
MDL Nummer: MFCD00468030
CAS nummer: 13739-02-1
EINECS: 237-310-2
Merck Index: 14,02960

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 241.0 to 245.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR