Dimethyltitanocene 5% in THF/toluene

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D4100-100G D4100-25G
TCIAD4100-100GEA 302 EUR
TCIAD4100-100G TCIAD4100-25G
Dimethyltitanocene 5% in THF/toluene
Dimethyltitanocene
Formule: C₁₂H₁₆Ti
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD02093882
CAS nummer: 1271-66-5
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fe32b7536a1340cd06eb97753580c39beeccaa33&vid=bd3253ce0928894f83971c7fb3c615f656677f00&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Orange to Red clear liquid
Concentration (Chelometric Titration) 5.0 to 5.5 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR