n-Dodecyl acrylate ≥98.0% (by GC) stabilised
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
Acrylic acid Lauryl ester, Acrylic acid Dodecyl ester, Dodecyl Acrylate, Lauryl Acrylate
Stabilisatie:
Stabilised with 4-methoxyphenol (MEHQ)
D4129-25ML
D4129-500ML
D4129-100ML
TCIAD4129-25MLEA
17.6
EUR
TCIAD4129-25ML
TCIAD4129-500ML
TCIAD4129-100ML
n-Dodecyl acrylate ≥98.0% (by GC) stabilised
n-Dodecyl acrylate
Formule:
C₁₅H₂₈O₂ MW: 240.387 g/mol Kookpunt: 120°C Smeltpunt: 4°C Vlampunt: 145°C |
MDL Nummer:
MFCD00042871 CAS nummer: 2156-97-0 EINECS: 218-463-4 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...