3,3'-Dihexyl-2,2'-bithiophene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D4180-1G D4180-5G
TCIAD4180-1GEA 150 EUR
TCIAD4180-1G TCIAD4180-5G
3,3'-Dihexyl-2,2'-bithiophene ≥98.0% (by GC)
3,3'-Dihexyl-2,2'-bithiophene
Formule: C₂₀H₃₀S₂
Kookpunt: 82°C
Bewaar temperatuur: Koelkast
CAS nummer: 125607-30-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR