4,4'-Dihexyl-2,2'-bithiophene ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D4182-200MG D4182-1G
TCIAD4182-200MGEA 75.9 EUR
TCIAD4182-200MG TCIAD4182-1G
4,4'-Dihexyl-2,2'-bithiophene ≥95.0% (by GC)
4,4'-Dihexyl-2,2'-bithiophene
Formule: C₂₀H₃₀S₂
Bewaar temperatuur: Koelkast
CAS nummer: 135926-94-2
Order Now
Specification Test Results
Purity (GC) min. 95.0 %
Specificaties
Environmentally Preferable