((2R,3R)-3-(Benzoyloxy)-4,4-difluoro-5-oxotetrahydrofuran-2-yl)methyl benzoate ≥98.0% (by HPLC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D4207-1G D4207-5G
TCIAD4207-1GEA 33 EUR
TCIAD4207-1G TCIAD4207-5G
((2R,3R)-3-(Benzoyloxy)-4,4-difluoro-5-oxotetrahydrofuran-2-yl)methyl benzoate ≥98.0% (by HPLC)
((2R,3R)-3-(Benzoyloxy)-4,4-difluoro-5-oxotetrahydrofuran-2-yl)methyl benzoate
Formule: C₁₉H₁₄F₂O₆
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD08458308
CAS nummer: 122111-01-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 121.0 to 125.0 deg-C
Specific rotation [a]20/D +69.0 to +73.0 deg (C=1, CH3CN)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR