2-(Diisopropylsilyl)pyridine ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: Diisopropyl(2-pyridyl)silane, PyDipSiH

D4264-1G D4264-5G
TCIAD4264-1GEA 79.2 EUR
TCIAD4264-1G TCIAD4264-5G
2-(Diisopropylsilyl)pyridine ≥95.0% (by GC)
2-(Diisopropylsilyl)pyridine
Formule: C₁₁H₁₉NSi
MW: 193.37 g/mol
Vlampunt: 84°C
MDL Nummer: MFCD22200515
CAS nummer: 1232692-92-0

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR