2,5-Dibromo-1,1,3,4-tetraphenylsilole ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D4310-1G
D4310-5G
TCIAD4310-1GEA
218
EUR
TCIAD4310-1G
TCIAD4310-5G
2,5-Dibromo-1,1,3,4-tetraphenylsilole ≥98.0% (by GC)
2,5-Dibromo-1,1,3,4-tetraphenylsilole
Formule:
C₂₈H₂₀Br₂Si |
CAS nummer:
200068-36-6 |
|
Specification Test Results
Purity (GC) | min. 98.0 % |
Melting point | 169.0 to 172.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...