4-(Difluoromethoxy)benzaldehyde ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D4334-1G D4334-5G
TCIAD4334-1GEA 23.1 EUR
TCIAD4334-1G TCIAD4334-5G
4-(Difluoromethoxy)benzaldehyde ≥98.0% (by GC)
4-(Difluoromethoxy)benzaldehyde
Formule: C₈H₆F₂O₂
Kookpunt: 87°C
Vlampunt: 109°C
MDL Nummer: MFCD00042252
CAS nummer: 73960-07-3
EINECS: 277-653-5
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %