5'-Deoxy-5-fluorocytidine ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D4342-1G D4342-5G
TCIAD4342-1GEA 17.6 EUR
TCIAD4342-1G TCIAD4342-5G
5'-Deoxy-5-fluorocytidine ≥98.0% (by HPLC, titration analysis)
5'-Deoxy-5-fluorocytidine
Formule: C₉H₁₂FN₃O₄
MDL Nummer: MFCD07369278
CAS nummer: 66335-38-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 203.0 to 210.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR