Lithium tetrachlorocuprate ∼2,5% ∼0.1 mol/L in THF

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D4395-100ML D4395-500ML
TCIAD4395-100MLEA 47.3 EUR
TCIAD4395-100ML TCIAD4395-500ML
Lithium tetrachlorocuprate ∼2,5% ∼0.1 mol/L in THF
Lithium tetrachlorocuprate
Formule: Cl₄CuLi₂
Vlampunt: -17°C
MDL Nummer: MFCD00011081
CAS nummer: 15489-27-7
UN: 2056
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a53fcfae3cefbeedfa6c28864be7c9d34ddf96ab&vid=8484ba37f7777b42929c77912e5ec65edb38cb66&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Orange to Amber to Dark red clear liquid
Concentration (Argentmetric titration) 2.2 to 2.9 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR