2-(Diphenylphosphino)benzaldehyde ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: (2-Formylphenyl)diphenylphosphine
D4411-1G D4411-5G
TCIAD4411-1GEA 28.6 EUR
TCIAD4411-1G TCIAD4411-5G
2-(Diphenylphosphino)benzaldehyde ≥97.0% (by GC)
2-(Diphenylphosphino)benzaldehyde
Formule: C₁₉H₁₅OP
MW: 290.30 g/mol
MDL Nummer: MFCD00013367
CAS nummer: 50777-76-9
Order Now
Specification Test Results
Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 115.0 to 119.0 deg-C