Diethyl (3-bromopropyl)phosphonate ≥95.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: (3-Bromopropyl)phosphonic Acid Diethyl Ester
D4434-1G D4434-5G
TCIAD4434-1GEA 33 EUR
TCIAD4434-1G TCIAD4434-5G
Diethyl (3-bromopropyl)phosphonate ≥95.0% (by GC)
Diethyl (3-bromopropyl)phosphonate
Formule: C₇H₁₆BrO₃P
MW: 259.08 g/mol
Kookpunt: 97°C
MDL Nummer: MFCD00800966
CAS nummer: 1186-10-3
Order Now
Specification Test Results
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 95.0 %