3,4-Dichlorobenzamide ≥98.0% (by HPLC, total nitrogen)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D4474-1G
D4474-5G
TCIAD4474-1GEA
46.2
EUR
TCIAD4474-1G
TCIAD4474-5G
3,4-Dichlorobenzamide ≥98.0% (by HPLC, total nitrogen)
3,4-Dichlorobenzamide
Formule:
C₇H₅Cl₂NO |
MDL Nummer:
MFCD00053009 CAS nummer: 2670-38-4 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 142.0 to 146.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...