2,4-Dichlorobenzyl bromide ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
1-(Bromomethyl)-2,4-dichlorobenzene
D4475-1G
D4475-5G
TCIAD4475-1GEA
36.3
EUR
TCIAD4475-1G
TCIAD4475-5G
2,4-Dichlorobenzyl bromide ≥98.0% (by GC)
2,4-Dichlorobenzyl bromide
Formule:
C₇H₅BrCl₂ Kookpunt: 112°C Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD03208523 CAS nummer: 20443-99-6 UN: 3261 ADR: 8,II |
Specification Test Results
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 30.0 to 34.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...