2,6-Dimethoxyaniline ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
D4480-1G
D4480-5G
TCIAD4480-1GEA
56.1
EUR
TCIAD4480-1G
TCIAD4480-5G
2,6-Dimethoxyaniline ≥98.0% (by GC)
2,6-Dimethoxyaniline
Formule:
C₈H₁₁NO₂ MW: 153.18 g/mol Kookpunt: 140°C |
MDL Nummer:
MFCD00053934 CAS nummer: 2734-70-5 |
Specification Test Results
Appearance | White to Gray to Brown powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 74.0 to 78.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...