(R)-(-)-DTBM-SEGPHOS® ≥99.0% (by HPLC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
(R)-(-)-4,4'-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-3,3'-bi(1,2-methylenedioxybenzene), (R)-(-)-DTBM-SEGPHOS®
D4501-1G
D4501-200MG
TCIAD4501-1GEA
125
EUR
TCIAD4501-1G
TCIAD4501-200MG
(R)-(-)-DTBM-SEGPHOS® ≥99.0% (by HPLC)
(R)-5,5'-Bis[di(3,5-di-tert-butyl-4-methoxyphenyl)phosphino]-4,4'-bi-1,3-benzodioxole
Formule:
C₇₄H₁₀₀O₈P₂ |
MDL Nummer:
MFCD09753003 CAS nummer: 566940-03-2 |
Specification Test Results
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 99.0 area% |
Optical purity (LC) | min. 99.0 ee% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...