3,5-Dimethyl-4-nitroisoxazole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D4506-5G D4506-25G
TCIAD4506-5GEA 37.4 EUR
TCIAD4506-5G TCIAD4506-25G
3,5-Dimethyl-4-nitroisoxazole ≥98.0% (by GC)
3,5-Dimethyl-4-nitroisoxazole
Formule: C₅H₆N₂O₃
MDL Nummer: MFCD00051656
CAS nummer: 1123-49-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 61.0 to 65.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR