1,2-Difluoro-4-(methylsulphonyl)benzene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 3,4-Difluorophenyl methyll sulphone

D4526-1G D4526-5G
TCIAD4526-1GEA 19.8 EUR
TCIAD4526-1G TCIAD4526-5G
1,2-Difluoro-4-(methylsulphonyl)benzene ≥98.0% (by GC)
1,2-Difluoro-4-(methylsulphonyl)benzene
Formule: C₇H₆F₂O₂S
MDL Nummer: MFCD04037934
CAS nummer: 424792-57-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 75.0 to 79.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR