2,7-Dibromo-9,9-diphenylfluorene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: 2,7-Dibromo-9,9-diphenyl-9H-fluorene

D4547-5G D4547-1G
TCIAD4547-5GEA 92.4 EUR
TCIAD4547-5G TCIAD4547-1G
2,7-Dibromo-9,9-diphenylfluorene ≥98.0% (by GC)
2,7-Dibromo-9,9-diphenylfluorene
Formule: C₂₅H₁₆Br₂
MW: 476.21 g/mol
MDL Nummer: MFCD09907958
CAS nummer: 186259-63-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e460b4219d6de71a8f27d0ff7fb990ba82bdde01&vid=e460b4219d6de71a8f27d0ff7fb990ba82bdde01&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR