Dimethylamine hydroiodide ≥98.0% (by total nitrogen and titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D4555-1G D4555-5G
TCIAD4555-1GEA 45.1 EUR
TCIAD4555-1G TCIAD4555-5G
Dimethylamine hydroiodide ≥98.0% (by total nitrogen and titration analysis)
Dimethylamine hydroiodide
Formule: C₂H₇N
CAS nummer: 51066-74-1
EINECS: 256-945-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 153.0 to 157.0 deg-C

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR