1,1,3,3,5,5,7,7,9,9,11,11-Dodecamethylhexasiloxane ≥96.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D4570-5ML
D4570-25ML
TCIAD4570-5MLEA
66
EUR
TCIAD4570-5ML
TCIAD4570-25ML
1,1,3,3,5,5,7,7,9,9,11,11-Dodecamethylhexasiloxane ≥96.0% (by GC)
1,1,3,3,5,5,7,7,9,9,11,11-Dodecamethylhexasiloxane
Formule:
C₁₂H₃₈O₅Si₆ Kookpunt: 74°C |
MDL Nummer:
MFCD00513634 CAS nummer: 995-82-4 |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...