2,3-Dibromonaphthalene ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 2,3-Diiodonaphthalene
D4597-200MG D4597-1G
TCIAD4597-200MGEA 52.8 EUR
TCIAD4597-200MG TCIAD4597-1G
2,3-Dibromonaphthalene ≥98.0% (by GC)
2,3-Dibromonaphthalene
Formule: C₁₀H₆Br₂
MDL Nummer: MFCD00021589
CAS nummer: 13214-70-5
Order Now
Specification Test Results
Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 140.0 to 144.0 deg-C