Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: [(Tetrahydropyran-2-yloxy)methyl]phosphonic acid Diethyl ester

D4611-200MG D4611-1G
TCIAD4611-200MGEA 41.8 EUR
TCIAD4611-200MG TCIAD4611-1G
Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate ≥96.0% (by GC)
Diethyl [(Tetrahydropyran-2-yloxy)methyl]phosphonate
Formule: C₁₀H₂₁O₅P
MW: 252.25 g/mol
Kookpunt: 95°C
MDL Nummer: MFCD09263515
CAS nummer: 71885-51-3

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR