3,5-Diethynylpyridine ≥96.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D4613-1G D4613-5G
TCIAD4613-1GEA 198 EUR
TCIAD4613-1G TCIAD4613-5G
3,5-Diethynylpyridine ≥96.0% (by GC)
3,5-Diethynylpyridine
Formule: C₉H₅N
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD11109669
CAS nummer: 67227-90-1

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 96.0 %
Melting point 70.0 to 74.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR