Diethyl-2-propynylmalonate ≥97.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D4616-1G D4616-5G
TCIAD4616-1GEA 114 EUR
TCIAD4616-1G TCIAD4616-5G
Diethyl-2-propynylmalonate ≥97.0% (by GC)
Diethyl-2-propynylmalonate
Formule: C₁₀H₁₄O₄
Kookpunt: 115°C
MDL Nummer: MFCD00091967
CAS nummer: 17920-23-9
Order Now
Specification Test Results
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %