2,5-Dibromo-1,1-dimethyl-3,4-diphenylsilole ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D4631-1G
D4631-5G
TCIAD4631-1GEA
218
EUR
TCIAD4631-1G
TCIAD4631-5G
2,5-Dibromo-1,1-dimethyl-3,4-diphenylsilole ≥98.0% (by GC)
2,5-Dibromo-1,1-dimethyl-3,4-diphenylsilole
Formule:
C₁₈H₁₆Br₂Si Bewaar temperatuur: Koelkast |
CAS nummer:
686290-22-2 |
|
Specification Test Results
Purity (GC) | min. 98.0 % |
Melting point | 161.0 to 165.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...