[1-[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]-4-iodobenzene](difluoroborane) ≥98.0% (by HPLC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: Boron, [2-[(3,5-dimethyl-2H-pyrrol-2-ylidene-κN)(4-iodophenyl)methyl]-3,5-dimethyl-1H-pyrrolato-κN]difluoro-, (T-4)-
D4632-200MG D4632-1G
TCIAD4632-200MGEA 119 EUR
TCIAD4632-200MG TCIAD4632-1G
[1-[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]-4-iodobenzene](difluoroborane) ≥98.0% (by HPLC)
[1-[(3,5-Dimethyl-1H-pyrrol-2-yl)(3,5-dimethyl-2H-pyrrol-2-ylidene)methyl]-4-iodobenzene](difluoroborane)
Formule: C₁₉H₁₈BF₂IN₂
MW: 450.08 g/mol
MDL Nummer: MFCD29905036
CAS nummer: 250734-47-5
Order Now
Specification Test Results
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 5.60 to 6.80 %
Lambda max. 500.0 to 505.0 nm (CH2Cl2)
Molar extinction coefficient min. 88000 (CH2Cl2, 500.0 to 505.0nm)