2,6-Diphenylbenzo[1,2-b:4,5-b']difuran

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

D4633-200MG D4633-1G
TCIAD4633-200MGEA 153 EUR
TCIAD4633-200MG TCIAD4633-1G
2,6-Diphenylbenzo[1,2-b:4,5-b']difuran
2,6-Diphenylbenzo[1,2-b:4,5-b']difuran
Formule: C₂₂H₁₄O₂
Smeltpunt: 345°C
CAS nummer: 5379-77-1

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Elemental analysis (Carbon) 82.50 to 87.50 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR