Dimethyltin oxide ≥95.0% (W)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D4649-25G D4649-100G
TCIAD4649-25GEA 74.8 EUR
TCIAD4649-25G TCIAD4649-100G
Dimethyltin oxide ≥95.0% (W)
Dimethyltin oxide
Formule: C₂H₆OSn
Smeltpunt: 395°C
MDL Nummer: MFCD00013838
CAS nummer: 2273-45-2
EINECS: 218-881-7
UN: 3146
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with ignition residue) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR