Decyltrimethoxysilane ≥97.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
D4704-25G
D4704-250G
TCIAD4704-25GEA
48.4
EUR
TCIAD4704-25G
TCIAD4704-250G
Decyltrimethoxysilane ≥97.0% (by GC)
Decyltrimethoxysilane
Formule:
C₁₃H₃₀O₃Si MW: 262.46 g/mol |
MDL Nummer:
MFCD00069135 CAS nummer: 5575-48-4 |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...