3,5-Dimethylisoxazole-4-carbonyl chloride ≥96.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Gevaar

D4706-1G D4706-5G
TCIAD4706-1GEA 47.3 EUR
TCIAD4706-1G TCIAD4706-5G
3,5-Dimethylisoxazole-4-carbonyl chloride ≥96.0% (by GC, titration analysis)
3,5-Dimethylisoxazole-4-carbonyl chloride
Formule: C₆H₆ClNO₂
Kookpunt: 90°C
MDL Nummer: MFCD00052555
CAS nummer: 31301-45-8
UN: 3265
ADR: 8,III

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %
Purity (Argentmetric Titration) min. 96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR