3',5'-Dichloro-2'-hydroxyacetophenone ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D4742-5G D4742-25G
TCIAD4742-5GEA 46.2 EUR
TCIAD4742-5G TCIAD4742-25G
3',5'-Dichloro-2'-hydroxyacetophenone ≥98.0% (by GC)
3',5'-Dichloro-2'-hydroxyacetophenone
Formule: C₈H₆Cl₂O₂
Kookpunt: 134°C
MDL Nummer: MFCD00016417
CAS nummer: 3321-92-4
Order Now
Specification Test Results
Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 95.0 to 99.0 deg-C