((Difluoromethyl)sulphonyl)benzene ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Difluoromethyl phenyl sulphone

D4764-1G D4764-5G
TCIAD4764-1GEA 36.3 EUR
TCIAD4764-1G TCIAD4764-5G
((Difluoromethyl)sulphonyl)benzene ≥98.0% (by GC)
((Difluoromethyl)sulphonyl)benzene
Formule: C₇H₆F₂O₂S
MDL Nummer: MFCD01050170
CAS nummer: 1535-65-5

Order Now

Specification Test Results

Appearance White or Colorless to Almost white or Almost colorless powder to lump to clear liquid
Purity (GC) min. 98.0 %
Refractive index n20/D 1.5000 to 1.5040


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR