3,6-Dibromo-9-(2-ethylhexyl)carbazole ≥98.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

D4830-5G D4830-1G
TCIAD4830-5GEA 295 EUR
TCIAD4830-5G TCIAD4830-1G
3,6-Dibromo-9-(2-ethylhexyl)carbazole ≥98.0% (by GC)
3,6-Dibromo-9-(2-ethylhexyl)carbazole
Formule: C₂₀H₂₃Br₂N
MDL Nummer: MFCD18252907
CAS nummer: 173063-52-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83c44a529b68b4b287b4aba8055c0f5887d404d4&vid=83c44a529b68b4b287b4aba8055c0f5887d404d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR