2-(Difluoromethoxy)benzaldehyde ≥98.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
D4837-1G D4837-5G
TCIAD4837-1GEA 31.9 EUR
TCIAD4837-1G TCIAD4837-5G
2-(Difluoromethoxy)benzaldehyde ≥98.0% (by GC)
2-(Difluoromethoxy)benzaldehyde
Formule: C₈H₆F₂O₂
Kookpunt: 83°C
Vlampunt: 107°C
MDL Nummer: MFCD00042251
CAS nummer: 71653-64-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e5907e39ae61772b43a0ca97ed47d89b234a83e&vid=4e5907e39ae61772b43a0ca97ed47d89b234a83e&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %