2,4-Dimethoxybenzaldehyde oxime ≥95.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Waarschuwing

Synoniemen: 2,4-Dimethoxybenzaldoxime

D4851-5G D4851-25G
TCIAD4851-5GEA 47.3 EUR
TCIAD4851-5G TCIAD4851-25G
2,4-Dimethoxybenzaldehyde oxime ≥95.0% (by GC)
2,4-Dimethoxybenzaldehyde oxime
Formule: C₉H₁₁NO₃
MW: 181.19
MDL Nummer: MFCD00082783
CAS nummer: 31874-34-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Melting point 105.0 to 109.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR