4-(Diethylamino)benzonitrile ≥98.0% (by GC, titration analysis)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
D4852-5G
D4852-25G
TCIAD4852-5GEA
41.8
EUR
TCIAD4852-5G
TCIAD4852-25G
4-(Diethylamino)benzonitrile ≥98.0% (by GC, titration analysis)
4-(Diethylamino)benzonitrile
Formule:
C₁₁H₁₄N₂ |
MDL Nummer:
MFCD00019754 CAS nummer: 2873-90-7 EINECS: 220-712-7 UN: 3439 ADR: 6.1,III |
|
Specification Test Results
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 66.0 to 70.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...