α,α-Difluoroanisole ≥98.0% (by GC)
Leverancier: TCI
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
D4914-5G
D4914-25G
TCIAD4914-5GEA
61.6
EUR
TCIAD4914-5G
TCIAD4914-25G
α,α-Difluoroanisole ≥98.0% (by GC)
α,α-Difluoroanisole
Formule:
C₇H₆F₂O Kookpunt: 140°C |
MDL Nummer:
MFCD00068184 CAS nummer: 458-92-4 EINECS: 207-283-1 UN: 1993 ADR: 3,III |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...