4-(Difluoromethoxy)toluene ≥96.0% (by GC)
Leverancier: TCI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Waarschuwing
Synoniemen: 1-(Difluoromethoxy)-4-methylbenzene
D4936-1G D4936-5G
TCIAD4936-1GEA 52.8 EUR
TCIAD4936-1G TCIAD4936-5G
4-(Difluoromethoxy)toluene ≥96.0% (by GC)
4-(Difluoromethoxy)toluene
Formule: C₈H₈F₂O
Kookpunt: 167°C
MDL Nummer: MFCD00042253
CAS nummer: 1583-83-1
EINECS: 216-434-0
Order Now
Specification Test Results
Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 96.0 %